missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10420-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene catalyzes the transport of phosphate into the mitochondrial matrix, either by proton cotransport or in exchange for hydroxyl ions. The protein contains three related segments arranged in tandem which are related to those found in other characterized members of the mitochondrial carrier family. Both the N-terminal and C-terminal regions of this protein protrude toward the cytosol.Specifications
SLC25A3 | |
Polyclonal | |
Unconjugated | |
SLC25A3 | |
OK/SW cl.48, PHC, Phosphate transport protein, PiC, PTP, SLC25A3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5250 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q00325 | |
SLC25A3 | |
SLC25A3 Fusion Protein Ag0667 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |