missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ SLC35A3 Monoclonal Antibody (4B6)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA521475

Product Code. 15559002

  • 4610.00 SEK / 100µg

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Description

Description

Peptide Sequence: DSKCSLRALN RVLHDEILNK PMETLKLAIP SGIYTLQNNL LYVALSNLDA ATY.

Uridine diphosphate-N-acetylglucosamine (UDP-GlcNAc) transporter in the Golgi apparatus. May supply UDP-GlcNAc as substrate for Golgi-resident glycosyltransferases that generate branching of diantennary oligosaccharides.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC35A3
Monoclonal
0.34 mg/mL
PBS with no preservative; pH 7.4
Q9Y2D2
SLC35A3
SLC35A3 (NP_036375, 61 a.a. ~ 113 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA
4B6
Unconjugated
SLC35A3
AMRS; golgi UDP-GlcNAc transporter; Slc35a3; solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member 3; solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member A3; solute carrier family 35 member 3; solute carrier family 35 member 3A; solute carrier family 35 member A3; UDP N-acetylglucosamine transporter; UDP-N-acetylglucosamine transporter
Mouse
Affinity chromatography
RUO
23443
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.