missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SLC39A7 Recombinant Protein (P01)
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH00645 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 7922 |
Molecular Weight (g/mol) | 73.7 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16112202
|
Abnova™
H00007922-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16122202
|
Abnova™
H00007922-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
AAH00645 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
73.7 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SLC39A7 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
7922 | |
SLC39A7 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DDLHDDLQEDFHGHSHRHSHEDFHHGHSHAHGHGHTHESIWHGHTHDHDHGHSHEDLHHGHSHGYSHESLYHRGHGHDHEHSHGGYGESGAPGIKQDLDAVTLWAYALGATVLISAAPFFVLFLIPVESNSPRHRSLLQILLSFASGGLLGDAFLHLIPHALEPHSHHTLEQPGHGHSHSGQGPILSVGLWVLSGIVAFLVVEKFVRHVKGGHGHSHGHGHAHSHTRGSHGHGRQERSTKEKQSSEEEEKETRGVQKRRGGSTVPKDGPVRPQNAEEEKRGLDLRVSGYLNLAADLAHNFTDGLAIGASFRGGRGLGILTTMTVLLHEVPHEVGDFAILVQSGCSKKQAMRLQLLTAVGALAGTACALLTEGGAVGSEIAGGAGPGWVLPFTAGGFIYVATVSVLPELLREASPLQSLLEVLGLLGGVIMMVLIAHLE | |
D6S115E/D6S2244E/H2-KE4/HKE4/KE4/RING5/ZIP7 | |
SLC39A7 | |
Wheat Germ (in vitro) | |
GST |