missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC44A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SLC44A1 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16823454
|
Proteintech
14687-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16813454
|
Proteintech
14687-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SLC44A1 encodes a choline transporter and may be involved in membrane synthesis and myelin production.Specifications
SLC44A1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q6X893, Q8VII6, Q8WWI5 | |
100434, 23446, 85254 | |
SLC44A1 Fusion Protein Ag6024 | |
Primary | |
-20°C | |
SLC44A1 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD92, CDW92, choline transporter-like protein 1, CHTL1, CTL1, SLC44A1 | |
Slc44a1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |