missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550624
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total SLC6A4 / 5-HTT protein.
SLC6A4 Polyclonal antibody specifically detects SLC6A4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
SLC6A4 | |
Polyclonal | |
Unconjugated | |
SLC6A4 | |
5 OCD1; 5HT transporter; 5HTT; 5-HTT; 5-HTTLPR; 5-hydroxytryptamine (serotonin) transporter; AI323329; HGNC:11050; hSERT; HTT; Na+/Cl- dependent serotonin transporter; OCD1; OTTHUMP00000163633; serotonin transporter; serotonin transporter 1; Sert; SERT1; Slc6a4; Sodium-dependent serotonin transporter; Sodium-dependent serotonin transporter (5HT transporter) (5HTT); solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); Solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); solute carrier family 6 (neurotransmitter transporter), member 4; solute carrier family 6 (neurotransmitter transporter, serotonin), member 4; solute carrier family 6 member 4; solute carrier family 6, member 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15567, 25553, 6532 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P31645, P31652, Q60857 | |
SLC6A4 | |
Synthetic peptide corresponding to a region derived from internal residues of human solute carrier family 6 (neurotransmitter transporter), member 4 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |