missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC7A8 Monoclonal Antibody (OTI7D6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA524944
Additional Details : Weight : 0.01000kg
Description
SLC7A8 Monoclonal antibody specifically detects SLC7A8 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
SLC7A8 | |
Monoclonal | |
0.74 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UHI5 | |
SLC7A8 | |
Full-length protein expressed in 293T cell transfected with human SLC7A8 expression vector | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7D6 | |
Unconjugated | |
SLC7A8 | |
4F2-LC5; 4F2LC-5; AA408822; cationic amino acid transporter y+ system; glycoprotein-associated amino acid transporter LAT2; hLAT2; integral membrane protein E16H; Large neutral amino acids transporter small subunit 2; LAT2; Lat4; LPI-PC1; L-type amino acid transporter 2; mLAT2; Slc7a8; solute carrier family 7 (amino acid transporter light chain, L system), member 8; solute carrier family 7 (amino acid transporter, L-type), member 8; solute carrier family 7 (cationic amino acid transporter, y+ system), member 8; solute carrier family 7 member 8; solute carrier family 8 (cationic amino acid transporter, y+ system), member 7 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23428 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |