missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC9A11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568334
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Human, Mouse, and Rat samples.
SLC9A11 is involved in pH regulation.Specifications
SLC9A11 | |
Polyclonal | |
Unconjugated | |
SLC9C2 | |
SLC9C2 | |
Synthesized peptide derived from human SLC9A11. | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5TAH2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
284525, 364029 | |
-20°C | |
Liquid |