missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLFNL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17065-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
SLFNL1 belongs to the Schlafen family. The function of the SLFNL1 protein remains unknown. There are four different isoforms.Specifications
SLFNL1 | |
Polyclonal | |
Unconjugated | |
SLFNL1 | |
schlafen like 1, Schlafen like protein 1, SLFNL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
200172 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q499Z3 | |
SLFNL1 | |
SLFNL1 Fusion Protein Ag10887 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |