Learn More
Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassette Accessories
Load samples into Slide-A-Lyzer dialysis cassettes and float them in dialysate buffer using these syringes and buoys.
Brand:  Thermo Scientific™ 66431
Additional Details : Weight : 1.81440kg
 View more versions of this product
Product Code. 10026323
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Description
Description
These accessories facilitate use of Slide-A-Lyzer Dialysis Cassettes. Three different sizes of syringes with 18-gauge beveled-tip needles enable efficient injection of samples into the various sizes of cassettes. The syringes also enable accurate and complete recovery of samples following dialysis.
Three flotation buoys enable Slide-A-Lyzer Dialysis Casettes to be suspended in buffer or other dialysate. The reusable buoys are made of low density polyethylene (LDPE). For the 0.5 and 3mL cassette sizes, single-cassette and 10-cassette buoys are offered. The 12mL cassette is wider than the other two sizes and requires its own model of buoy. The 30mL Slide-A-Lyzer Dialysis Cassettes contain their own air chamber and do not require use of a float buoy.
Highlights:
  • Reusable – polypropylene syringes with stainless steal needles and the LDPE float buoys are easily washed and reused many times
  • Complete – these are the only two kinds of special accessories one needs for trouble-free use of Slide-A-Lyzer Dialysis Cassettes
Item-Specific Features:
  • Single buoys (Part No. 66430) each hold one 0.1–0.5mL or 0.5–3mL cassette
  • Single buoys (Part No. 66432) each hold one 3–12mL capacity cassette
  • Carousel Buoys (Part No. 66431) each hold ten 0.1–0.5mL or 0.5–3mL cassettes
  • Needles supplied with the syringes are sterile, 1' long
Includes:
Each syringe pack contains 10 syringes and 10 needles.
Specifications
Specifications
Slide-A-Lyzer Cassette Carousel Float Buoy (for 0.5mL to 3mL cassettes)
Low density polyethylene (LDPE)
1 buoy
Floating ten 0.5mL or 3mL Cassettes simultaneously
Videos