Learn More
Thermo Scientific™ Sliding Drawer Racks for Tubes (4 inner door freezers)
Maximize tube storage capacity per freezer with a racking solution for four inner-door, ultra-low temperature freezers.
Brand:  Thermo Scientific™ 1950801
Additional Details : Weight : 2.30000kg
 View more versions of this product
Product Code. 15160529
  • 1276.44 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
Thermo Scientific freezer racking and tubing solutions offer a convenient, streamlined process to maximize sample storage in your laboratory. Our racking solutions are designed to hold tubes ranging from 200μL up to 5.0mL, offering maximized sample storage capacity for both five and four inner door freezers.
Specifications
Specifications
14 cm
4
Tubes
16
30.2 cm
EXF and HFU 200, 300, 400 and 600 Series
42.2 cm
4
48/Box
Sliding Drawer (Tubes)