Läs mer
Edmund Buhler Universal Shaker SM 30 C
Beskrivning
- The shaking motion of the laboratory shaker SM 30 C can be switched from orbital to horizontal
- This guarantees flexibility for different applications with one device
- The universal shaker is ideally suited for high loads up to 30 kg and for mixing difficult media
- The wide choice of rack systems ensures optimal fastening of different vessels
Specifikationer
Specifikationer
| Hastighetsintervall | 15 till 300 rpm |
| Elektriska krav | 115/230 V, 50/60 Hz |
| Spänning | 115/230 V |
| Temperaturområde (metriskt) | 5 °C till 50 °C |
| Frekvens | 50/60 Hz |
| Djup (metriskt) | 610 mm |
| Bredd (metrisk) | 680 mm |
| Fraktvikt (metrisk) | 33 kg |
| Modell | SM 30 C |
| Fuktighetsområde | 85% |
| Visa mer |