Learn More
SMAD2 Mouse anti-Human, Unconjugated, Clone: 2H10C7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67343-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16849925
  • 1656.73 SEK / 20µL
Estimated Shipment: 31-05-2024
to see stock.
Description
Description
The SMAD2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SMAD2. This antibody recognizes human, mouse, rat antigen. The SMAD2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SMAD2
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
2H10C7
Unconjugated
20 μL
4087
IgG1