missing translation for 'onlineSavingsMsg'
Learn More

SMAD9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16397-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826014

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the SMAD family, which transduces signals from TGF-beta family members. The encoded protein is activated by bone morphogenetic proteins and interacts with SMAD4. Two transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

SMAD9
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15198, O54835, Q9JIW5
MAD homolog 9, MADH6, MADH9, Mothers against DPP homolog 9, SMAD 9, SMAD family member 9, SMAD8A, SMAD8B, SMAD9
Rabbit
IgG
20 μL
-20°C
Primary
4093, 55994, 85435
Western Blot
Unconjugated
SMAD9
Liquid
SMAD9
SMAD9 Fusion Protein Ag9490
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Mouse, Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers