missing translation for 'onlineSavingsMsg'
Learn More

SMARCA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26613-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16875025

  • 4721.67 SEK / 150µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SMARCA2 encodes a protein that is a member of the SWI/SNF family of proteins and is highly similar to the brahma protein of Drosophila. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. Two transcript variants encoding different isoforms have been found for this gene, which contains a trinucleotide repeat (CAG) length polymorphism.
Specifications

Specifications

SMARCA2
Polyclonal
Unconjugated
SMARCA2
ATP dependent helicase SMARCA2, BAF190, BAF190B, BRG1 associated factor 190B, BRM, hBRM, hSNF2a, Protein brahma homolog, SMARCA2, SMARCA2-Specific, SNF2, SNF2 alpha, SNF2A, SNF2L2, SNF2LA, Sth1p, SWI2
Rabbit
Antigen Affinity Chromatography
RUO
6595, 67155
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P51531, Q6DIC0
SMARCA2
SMARCA2-Specific Fusion Protein Ag25157
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers