Learn More
Moldex™ Smart FFP-Mask
Product Code. 12352299
Change view
Click to view available options
For Use With (Application):
For Protection Against Fine Toxic Dust, Oil & Water Based Mists and Fumes
Nuisance Relief or Protection Against Ozone
Unit Size:
Pack of 10
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
12352299 For Protection Against Fine Toxic Dust, Oil & Water Based Mists and Fumes Pack of 20
12392299 Nuisance Relief or Protection Against Ozone Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12352299 Supplier Moldex™ Supplier No. 248001
  • ONLINE EXCLUSIVE PRICE
    1338.27 SEK
    544.07 SEK / Paket med 20
    Save 794.20 SEK
    59% Av
I lager
Add to basket
Add to basket
Protects against Ozone, and nuisance relief
Moldex Smart Series FFP2 NR D non-valved dust mask.
Suited to most industrial applications this general purpose mask is strong and durable. The ActiveForm™ seal automatically adjusts to different face shapes and movements to give a secure seal.
Specifications
Certifications/Compliance EN 149:2001+A1:2009
Disposable Single-use
Product Type Smart FFP-Mask
For Use With (Application) For Protection Against Fine Toxic Dust, Oil & Water Based Mists and Fumes