Learn More
Moldex™ Smart FFP-Mask
Protects against Ozone, and nuisance relief
1060.00 SEK - 1530.00 SEK
Specifications
Certifications/Compliance | EN 149:2001+A1:2009 |
---|---|
Disposable | Single-use |
Product Type | Smart FFP-Mask |
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
12352299
|
Moldex™
248001 |
For Protection Against Fine Toxic Dust, Oil & Water Based Mists and Fumes |
1060.00 SEK
Pack of 20 |
In Stock
Log in to see stock. |
|||||
12392299
|
Moldex™
244501 |
Nuisance Relief or Protection Against Ozone |
1530.00 SEK
Pack of 10 |
N/A | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
Moldex Smart Special FFP2 NR D + Ozone 10 x WEL valved dust mask.The activated layer of charcoal built into this mask offers relief from low levels of gases and vapours, often found in conjunction with particulate hazards. FFP2 NR D + Ozone 10 x WEL.
Specifications
EN 149:2001+A1:2009 | |
Single-use | |
Smart FFP-Mask |