Learn More
Smart SolutionsTM TerpiMix Kit 2500 μg/ml i isopropanol, Dr. Ehrenstorfer
Product Code. 30317000
Change view
Click to view available options
Kvantitet:
3 x 1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30317000 3 x 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317000 Supplier Dr. Ehrenstorfer Supplier No. DREK50000655IP
 Begränsad artikel
Beräknad leverans: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Upptäck Dr. Ehrenstorfers certifierade referensmaterial: finns tillgängliga i flera format, inklusive flerkomponents regulatoriska blandningar, för att driva din livsmedels- och miljöanalys med spårbar, ISO-ackrediterad kvalitet
TRUSTED_SUSTAINABILITY
Spécification
Kemiskt namn eller material (-)-alpha-Cedrene∣(-)-Borneol∣(-)-Fenchone∣(-)-Isopulegol∣(-)-Trans-Caryophyllene∣(-)-Verbenone∣(+)-Borneol∣(+)-Carvone∣(+)-Dihydrocarveol∣(+)-Fenchone∣(+)-Fenchyl Alcohol∣(±)-beta-Citronellol∣(1S)-(+)-3-Carene∣(cis + trans)-Nerolidol∣1-Methyl-2-isopropylbenzene∣1-Terpinen-4-ol∣3-Isopropyltoluene (m-Cymol)∣4-Cymene∣4-Thujanol∣Acetic Acid-Octyl Ester∣alpha-(-)-Thujone∣alpha-Bisabolol∣alpha-Farnesene∣alpha-Humulene∣alpha-Phellandrene∣alpha-Terpinene∣alpha-Terpineol∣beta-Ocimene∣beta-Pinene∣Camphene∣Carvacrol∣Caryophyllene Oxide∣Cedrol∣Citral∣Delta-3-Carene∣delta-Valerolactam∣DL-alpha-Pinene∣DL-Camphor∣DL-Linalool∣Eucalyptol (1,8-Cineole)∣Farnesol (Mixture of Isomers)∣gamma-Terpinene∣Geraniol∣Geranyl Acetate∣Guaiol∣Isoborneol∣Isobornyl Acetate∣Limonene∣Menthol Racemic∣Menthone∣Myrcene∣Nerol∣Nootkatone∣Phytane∣Phytol (cis/trans mixture)∣Piperitone∣R-(+)-Pulegone∣Sabinene∣Safranal∣Squalene∣Terpinolene∣Thymol∣Valencene
CAS 469-61-4, 464-45-9, 7787-20-4, 89-79-2, 87-44-5, 1196-01-6
Analyt- eller komponentnamn (-)-alpha-Cedrene∣(-)-Borneol∣(-)-Fenchone∣(-)-Isopulegol∣(-)-Trans-Caryophyllene∣(-)-Verbenone∣(+)-Borneol∣(+)-Carvone∣(+)-Dihydrocarveol∣(+)-Fenchone∣(+)-Fenchyl Alcohol∣(±)-beta-Citronellol∣(1S)-(+)-3-Carene∣(cis + trans)-Nerolidol∣1-Methyl-2-isopropylbenzene∣1-Terpinen-4-ol∣3-Isopropyltoluene (m-Cymol)∣4-Cymene∣4-Thujanol∣Acetic Acid-Octyl Ester∣alpha-(-)-Thujone∣alpha-Bisabolol∣alpha-Farnesene∣alpha-Humulene∣alpha-Phellandrene∣alpha-Terpinene∣alpha-Terpineol∣beta-Ocimene∣beta-Pinene∣Camphene∣Carvacrol∣Caryophyllene Oxide∣Cedrol∣Citral∣Delta-3-Carene∣delta-Valerolactam∣DL-alpha-Pinene∣DL-Camphor∣DL-Linalool∣Eucalyptol (1,8-Cineole)∣Farnesol (Mixture of Isomers)∣gamma-Terpinene∣Geraniol∣Geranyl Acetate∣Guaiol∣Isoborneol∣Isobornyl Acetate∣Limonene∣Menthol Racemic∣Menthone∣Myrcene∣Nerol∣Nootkatone∣Phytane∣Phytol (cis/trans mixture)∣Piperitone∣R-(+)-Pulegone∣Sabinene∣Safranal∣Squalene∣Terpinolene∣Thymol∣Valencene
Frakt skick Rumstemperatur
Certifikat och efterlevnad ISO 17034
Lösningsmedel eller matris Isopropylalkohol
Kvantitet 3 x 1 ml
Koncentration eller sammansättning (efter analyt eller komponenter) (-)-alpha-Cedrene 2500 μg/mL∣(-)-Borneol 2500 μg/mL∣(-)-Fenchone 2500 μg/mL∣(-)-Isopulegol 2500 μg/mL∣(-)-Trans-Caryophyllene 2500 μg/mL∣(-)-Verbenone 2500 μg/mL∣(+)-Borneol 2500 μg/mL∣(+)-Carvone 2500 μg/mL∣(+)-Dihydrocarveol 2500 μg/mL∣(+)-Fenchone 2500 μg/mL∣(+)-Fenchyl Alcohol 2500 μg/mL∣(±)-beta-Citronellol 2500 μg/mL∣(1S)-(+)-3-Carene 2500 μg/mL∣(cis + trans)-Nerolidol 2500 μg/mL∣1-Methyl-2-isopropylbenzene 2500 μg/mL∣1-Terpinen-4-ol 2500 μg/mL∣3-Isopropyltoluene (m-Cymol) 2500 μg/mL∣4-Cymene 2500 μg/mL∣4-Thujanol 2500 μg/mL∣Acetic Acid-Octyl Ester 2500 μg/mL∣alpha-(-)-Thujone 2500 μg/mL∣alpha-Bisabolol 2500 μg/mL∣alpha-Farnesene 2500 μg/mL∣alpha-Humulene 2500 μg/mL∣alpha-Phellandrene 2500 μg/mL∣alpha-Terpinene 2500 μg/mL∣alpha-Terpineol 2500 μg/mL∣beta-Ocimene 2500 μg/mL∣beta-Pinene 2500 μg/mL∣Camphene 2500 μg/mL∣Carvacrol 2500 μg/mL∣Caryophyllene Oxide 2500 μg/mL∣Cedrol 2500 μg/mL∣Citral 2500 μg/mL∣Delta-3-Carene 2500 μg/mL∣delta-Valerolactam 2500 μg/mL∣DL-alpha-Pinene 2500 μg/mL∣DL-Camphor 2500 μg/mL∣DL-Linalool 2500 μg/mL∣Eucalyptol (1,8-Cineole) 2500 μg/mL∣Farnesol (Mixture of Isomers) 2500 μg/mL∣gamma-Terpinene 2500 μg/mL∣Geraniol 2500 μg/mL∣Geranyl Acetate 2500 μg/mL∣Guaiol 2500 μg/mL∣Isoborneol 2500 μg/mL∣Isobornyl Acetate 2500 μg/mL∣Limonene 2500 μg/mL∣Menthol Racemic 2500 μg/mL∣Menthone 2500 μg/mL∣Myrcene 2500 μg/mL∣Nerol 2500 μg/mL∣Nootkatone 2500 μg/mL∣Phytane 2500 μg/mL∣Phytol (cis/trans mixture) 2500 μg/mL∣Piperitone 2500 μg/mL∣R-(+)-Pulegone 2500 μg/mL∣Sabinene 2500 μg/mL∣Safranal 2500 μg/mL∣Squalene 2500 μg/mL∣Terpinolene 2500 μg/mL∣Thymol 2500 μg/mL∣Valencene 2500 μg/mL
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.