Learn More
Thermo Scientific™ Smart-Vue Pro Alert Software
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you.
POA
Specifications
Type | Smart-Vue Pro Software and Subscription Service |
---|---|
Certifications/Compliance | Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance |
For Use With (Equipment) | Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module |
Description
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions, Smart-Vue™ Alert will automatically notify the user based notification preferences wherever you are (phone, text or email).
- Add-on notification package through SMS and voice calls (Twillo)
- 12 month license (post-activation)
- License depends on the number of monitoring points desired
Specifications
Smart-Vue Pro Software and Subscription Service | |
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance | |
Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module |