Learn More
Thermo Scientific™ Smart-Vue Pro Modules
Increase efficiency with multi-sensor wireless monitoring
POA
Specifications
Type Module
Certifications/Compliance Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
View More Specs
Description
Description
Smart-Vue Pro modules are available in two configurations- Duo and Quatro. The Duo allows up to 2 sensor channels for monitoring. The Quatro has extended range of 4 sensor channels for 2 wired and 2 wireless sensors. Ideal for monitoring several pieces of equipment simultaneously or multiple points within one piece of equipment
Bright 2.4 in. color touch screen with multiple views Alarms locally with real -time audio-visual alarms on the modules Audio visual alarms for local notification Communication locally via Bluetooth and to the system via LoRaLow AC power consumption through USB port access Optional replaceable battery power with extended life (minimum of 1 year) allows continuous monitoring Choice of wired and wireless sensors with automatic sensor detection User traceability with customized levels of user access with encrypted password protection
Specifications
Specifications
Module
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance