Learn More
Thermo Scientific™ Smart-Vue Pro On-Premise Software and Subscription Services
A simplified software experience, allowing you to focus on what matters the most
Brand:  Thermo Scientific™ SVPSWSM21500
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 16612207
  • 619665.00 SEK / Each
Estimated Shipment: 21-09-2022
to see stock.
Add to basket
Description
Description
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions the system will automatically notify the user based on prescribed notification preferences and allows you to document notes around each specific event for audit purposes. The floorplan view and local audio visual alarms will make it easy to locate alarming equipment during the critical recovery times. The on-premise solution maintains your Smart-Vue Pro data locally on your site's server.
  • Intuitive dashboard simplifies daily usage.
  • Optional floor plan and sites views.
  • Data can be exported with a click for reporting and customizable graphs.
  • Web application on Cloud or customer server.
  • Instant access to alarms, readings, and graphs.
  • Remote data access.
  • Flexible organization by site, department user, equipment, and criticality.
  • And trail and secure-level access control to support 21CFR Part 11
  • Cloud-based configurations allow flexibility to check status anytime, anywhere from any modern web browser.
Specifications
Specifications
Smart-Vue Server maintenance contract after the first year to access software update and upgrade - validity 1 year; Up to 1500 PTS
Smart-Vue Pro System
Smart-Vue Pro Software and Subscription Service
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance