Learn More
LevGo™ SmartSlicer™ Safety Razor Blades
Say no to dangerous sharps. SmartSlicer™ Safety Razor Blades won't scratch surfaces when cutting agarose gels.
389.00 SEK - 417.00 SEK
Specifications
Blade Material | Food Grade PET |
---|---|
Product Type | Blade |
For Use With (Application) | Slicing Agarose Gels, Cutting Soft Tissue and Plant Matter |
Disposable | Single-use |
Sterility | Non-sterile |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15326558
|
LevGo™
HS120260 |
Large |
389.00 SEK
Pack of 100 |
Estimated Shipment: 27-08-2025
Log in to see stock. |
||||||
15316558
|
LevGo™
HS120259 |
Small |
417.00 SEK
Pack of 100 |
In Stock
Log in to see stock. |
||||||
Description
- Cleanly slice agarose gels* right on the light box without scratching the surface.
- Ideal for cutting soft tissue and plant matter.
- Adjust the size required by simply cutting with scissors.
- Available in two sizes and made in the USA from food grade PET.
- Supplied in packs of 100.
Specifications
Food Grade PET | |
Slicing Agarose Gels, Cutting Soft Tissue and Plant Matter | |
Non-sterile | |
Food Grade | |
No |
Blade | |
Single-use | |
0.22 mm | |
100 Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
LevGo™ SmartSlicer™ Safety Razor Blades