missing translation for 'onlineSavingsMsg'
Learn More

SMCHD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25589-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16874145

  • 4721.67 SEK / 150µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein which contains a hinge region domain found in members of the SMC (structural maintece of chromosomes) protein family.
Specifications

Specifications

SMCHD1
Polyclonal
Unconjugated
SMCHD1
KIAA0650, SMCHD1
Rabbit
Antigen Affinity Chromatography
RUO
23347, 74355
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A6NHR9, Q6P5D8
Smchd1
SMCHD1 Fusion Protein Ag22293
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers