Learn More
Sartorius™ Smooth Surface Filter Discs Qualitative Filter Papers, Grade: 3 w
Used for analytical and technical purposes.
Brand:  Sartorius™ FT-3-308-090
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10082371
  • 134.00 SEK / Pack of 100
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
Description
Description
  • For routine analyses like clarification, determination of substances
  • Discs with a center hole for technical applications
  • Made of refined pulp and cotton linters with >95% α-cellulose content
  • Smooth surface
  • Wet-strengthened
Specifications
Specifications
Disc
15μm/sec.
3 w
0.14 mm
90 mm
Cotton Linters, Refined Pulp
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
100 Pack
Qualitative Filter Paper
90 mm