Learn More
Alfa Aesar™ Smopex™-234, metal scavenger
Brand:  Alfa Aesar™ 044984.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15462087
  • 923.65 SEK / 5g
Estimated Shipment date
from Supplier 30-01-2018
Add to basket
Description
Description
  • Recovery of precious metals from catalyst processes, especially Pd, Rh, Ru, Pt, and Ir
  • Also, base metals removed: Sn
Specifications
Chemical Identifiers
Smopex™-234 metal scavenger
912338-63-7
Mercaptoethylacrylate grafted polyolefin fiber
Specifications
Smopex™-234 metal scavenger
Powder
Off-white
912338-63-7
Mercaptoethylacrylate grafted polyolefin fiber
5g