missing translation for 'onlineSavingsMsg'
Learn More

SMOX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15052-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824034

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Polyamines are ubiquitous polycationic alkylamines which include spermine, spermidine, putrescine, and agmatine. These molecules participate in a broad range of cellular functions which include cell cycle modulation, scavenging reactive oxygen species, and the control of gene expression. These molecules also play important roles in neurotransmission through their regulation of cell-surface receptor activity, involvement in intracellular signalling pathways, and their putative roles as neurotransmitters. This gene encodes an FAD-containing enzyme that catalyzes the oxidation of spermine to spermadine and secondarily produces hydrogen peroxide. Multiple transcript variants encoding different isoenzymes have been identified for this gene, some of which have failed to demonstrate significant oxidase activity on natural polyamine substrates.
Specifications

Specifications

SMOX
0.47 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99K82, Q9NWM0
C20orf16, PAO, PAO 1, PAOh1, Polyamine oxidase 1, SMO, SMOX, spermine oxidase
Rabbit
IgG
20 μL
-20°C
Primary
228608, 308652, 54498
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SMOX
Liquid
SMOX
SMOX Fusion Protein Ag7035
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers