missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SMPDL3B (Human) Recombinant Protein

Human SMPDL3B full-length ORF ( NP_001009568.1, 1 a.a. - 373 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00027293-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138742

  • 4515.00 SEK / 10µg
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MRLLAWLIFLANWGGARAEPGKFWHIADLHLDPDYKVSKDPFQVCPSAGSQPVPDAGPWGDYLCDSPWALINSSIYAMKEIEPEPDFILWTGDDTPHVPDEKLGEAAVLEIVERLTKLIREVFPDTKVYAALGNHDFHPKNQFPAGSNNIYNQIAELWKPWLSNESIALFKKGAFYCEKLPGPSGAGRIVVLNTNLYYTSNALTADMADPGQQFQWLEDVLTDASKAGDMVYIVGHVPPGFFEKTQNKAWFREGFNEKYLKVVRKHHRVIAGQFFGHHHTDSFRMLYDDAGVPISAMFITPGVTPWKTTLPGVVNGANNPAIRVFEYDRATLSLKVRSPAEARGGGWEGLKCITTFPHSQLIHLPLTTEPQEG
Specifications

Specifications

NP_001009568.1
sphingomyelin phosphodiesterase, acid-like 3B
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SMPDL3B
GST
27293
Wheat germ expression system
10 μg
ASML3B
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers