missing translation for 'onlineSavingsMsg'
Learn More

SMPX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15791-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16895124

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SMPX plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair.
Specifications

Specifications

SMPX
Polyclonal
Unconjugated
SMPX
small muscle protein, X linked, Small muscular protein, SMPX, SRMX
Rabbit
Antigen Affinity Chromatography
RUO
23676, 66106, 84416
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q925F0, Q9DC77, Q9UHP9
Smpx
SMPX Fusion Protein Ag8497
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers