missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SMR3B Recombinant Protein

Brand:  Abnova™ H00010879-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16115792

  • 6150.00 SEK / 25µg
Estimated Shipment: 07-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH15327
Solution
10879
SMR3B (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SMR3B
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
34.3
8
Glutathione Sepharose 4 Fast Flow
25 ug
MKSLTWILGLWALAACFTPGESQRGPRGPYPPGPLAPPQPFGPGFVPPPPPPPYGPGRIPPPPPAPYGPGIFPPPPPQP
MGC104379/P-B/PBII/PRL3/PROL3/SMR1B
SMR3B
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers