Learn More
SMYD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16151-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Acts as a transcriptional repressor. Essential for cardiomyocyte differentiation and cardiac morphogenesis.Specifications
SMYD1 | |
Polyclonal | |
Unconjugated | |
SMYD1 | |
BOP, SMYD1, ZMYND18, ZMYND22 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12180, 150572 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97443, Q8NB12 | |
SMYD1 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |