Läs mer
SN-38 Glucuronide, TRC
Produktkod. 30409074
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30409074 10 mg
1 options
Produktkod. 30409074 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS58998010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 121080-63-5
Molekylformel C28 H28 N2 O11
Molekylvikt (g/mol) 568.53
Synonym β-D-Glucopyranosiduronic acid, (4S)-4,11-diethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl, β-D-Glucopyranosiduronic acid, 4,11-diethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl, (S)-, (4S)-4,11-Diethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl β-D-glucopyranosiduronic acid, SN 38G, SN-38 glucuronide, 10-Hydroxy-7-ethylcamptothecin 10-β-D-glucuronide, 7-Ethyl-10-hydroxy-20(S)-camptothecin 10-β-D-glucuronide, 7-Ethyl-10-hydroxycamptothecin 10-β-D-glucuronide, (4S)-4,11-Diethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione 9-(β-D-glucopyranosiduronide), 11-Ethyl-9-hydroxycamptothecin 9-β-D-glucuronide
LEDER CCc1c2CN3C(=O)C4=C(C=C3c2nc5ccc(O[C@@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O)C(=O)O)cc15)[C@@](O)(CC)C(=O)OC4
Specifikationer
Kemiskt namn eller material SN-38 Glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C28H28N2O11/c1-3-12-13-7-11(40-26-22(33)20(31)21(32)23(41-26)25(35)36)5-6-17(13)29-19-14(12)9-30-18(19)8-16-15(24(30)34)10-39-27(37)28(16,38)4-2/h5-8,20-23,26,31-33,38H,3-4,9-10H2,1-2H3,(H,35,36)/t20-,21-,22+,23-,26+,28-/m0/s1
Formel vikt 568.1693
Procent renhet >95%