Läs mer
SN-38 Tri-O-acetyl-Beta-D-glucuronic Acid Methyl Ester, TRC
Produktkod. 30419593
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30419593 5 mg
1 options
Produktkod. 30419593 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS5899955MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-4,11-Diethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl beta-D-Glucopyranosiduronic Acid Methyl Ester 2,3,4-Triacetate
CAS 121098-77-9
Molekylformel C35H36N2O14
Kvantitet 5 mg
Synonym SN-38 Tri-O-acetyl-beta-D-glucuronic Acid Methyl Ester
InChI formel InChI=1S/C35H36N2O14/c1-7-19-20-11-18(50-33-30(49-17(5)40)28(48-16(4)39)27(47-15(3)38)29(51-33)32(42)45-6)9-10-24(20)36-26-21(19)13-37-25(26)12-23-22(31(37)41)14-46-34(43)35(23,44)8-2/h9-12,27-30,33,44H,7-8,13-14H2,1-6H3/t27-,28-,29-,30+,33+,35-/m0/s1
LEDER CCc1c2CN3C(=O)C4=C(C=C3c2nc5ccc(O[C@@H]6O[C@@H]([C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]6OC(=O)C)C(=O)OC)cc15)[C@@](O)(CC)C(=O)OC4
Molekylvikt (g/mol) 708.67
Formel vikt 708.2167