Läs mer
Tocris Bioscience™ SNAP
GREENER_CHOICE
Produktkod. 18045862
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18045862 10 mg
30001104 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18045862 Leverantör Tocris Bioscience™ Leverantörsnummer 0598/10
A stable analog of endogenous S-nitroso compounds
A stable analog of endogenous S-nitroso compounds. A source of NO in vivo which unlike organic O-nitrates does not induce tolerance. Decomposes slowly in solution with a t½ of 37h.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79032-48-7
Molekylformel C7H12N2O4S
Specifikationer
Kemiskt namn eller material SNAP
Kvantitet 10 mg
Formel vikt 220.24
Renhet >98%
Molekylvikt (g/mol) 220.24