missing translation for 'onlineSavingsMsg'
Learn More

SNAPC3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27062-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825355

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SNAPC3 is part of the SNAPc complex required for the transcription of both RNA polymerase II and III small-nuclear RNA genes. SNAPC3 binds to the proximal sequence element (PSE), a non-TATA-box basal promoter element common to these 2 types of genes. SNAPC3 recruits TBP and BRF2 to the U6 snRNA TATA box.
Specifications

Specifications

SNAPC3
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q92966
PTF subunit beta, PTFbeta, SNAP50, SNAPc 50 kDa subunit, SNAPc subunit 3, SNAPC3
Rabbit
IgG
20 μL
-20°C
Primary
6619
Western Blot
Unconjugated
SNAPC3
Liquid
SNAPC3
SNAPC3 Fusion Protein Ag19851
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers