missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAPC3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SNAPC3 |
---|---|
Concentration | 0.47 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825355
|
Proteintech
27062-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815355
|
Proteintech
27062-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SNAPC3 is part of the SNAPc complex required for the transcription of both RNA polymerase II and III small-nuclear RNA genes. SNAPC3 binds to the proximal sequence element (PSE), a non-TATA-box basal promoter element common to these 2 types of genes. SNAPC3 recruits TBP and BRF2 to the U6 snRNA TATA box.Specifications
SNAPC3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q92966 | |
6619 | |
SNAPC3 Fusion Protein Ag19851 | |
Primary | |
-20°C | |
SNAPC3 |
0.47 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PTF subunit beta, PTFbeta, SNAP50, SNAPc 50 kDa subunit, SNAPc subunit 3, SNAPC3 | |
SNAPC3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |