Learn More
SNAPC5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555068
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MLSRLQELRK EEETLLRLKA ALHDQLNRLK VEELALQSMI SSRRGDEMLS SHTVPEQSHD MLVHVDNEAS INQTTLELST KSHVTEEEEE EEEEESDS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.
SNAPC5 Polyclonal antibody specifically detects SNAPC5 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
SNAPC5 | |
Polyclonal | |
Unconjugated | |
SNAPC5 | |
2010103A03Rik; CTD-3185P2.1; small nuclear RNA activating complex polypeptide 5; small nuclear RNA activating complex, polypeptide 5; small nuclear RNA activating complex, polypeptide 5, 19kD; small nuclear RNA activating complex, polypeptide 5, 19kDa; small nuclear RNA-activating complex polypeptide 5; SNAP19; SNAPc 19 kDa subunit; SNAPc subunit 5; SNAPC5; snRNA-activating protein complex 19 kDa subunit; snRNA-activating protein complex subunit 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10302 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75971 | |
SNAPC5 | |
Recombinant protein corresponding to Human SNAPC5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only