missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNCB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10498-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Synuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer′s (AD), Parkinson′s PD) and breast cancer. In AD, a peptide derived from alpha synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of alpha synuclein has been linked to many familial cases of PD. In breast cancer increased expression of gamma synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.Specifications
SNCB | |
Polyclonal | |
Unconjugated | |
SNCB | |
Beta synuclein, SNCB, synuclein, beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104069, 113893, 6620 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q16143, Q63754, Q91ZZ3 | |
SNCB | |
SNCB Fusion Protein Ag0772 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |