Learn More
SND1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540124
Additional Details : Weight : 0.00910kg
Description
Recommended positive controls: 293T and HepG2 cells. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (89%), Xenopus laevis (93%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SND1 Polyclonal antibody specifically detects SND1 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
SND1 | |
Polyclonal | |
Unconjugated | |
Snd1 | |
100 kDa coactivator; 4SNc-Tudor domain protein; 4SNc-Tudor domain protein long form; 4SNc-Tudor domain protein short form; AL033314; EBNA-2 co-activator (100kD); EBNA2 coactivator p100; p100; p100 co-activator; p100 co-activator variant 1; p100 EBNA2 co-activator; p100L; P100-like protein short variant; p100S; similar to Homo sapiens EBNA-2 co-activator (100kDa coactivator); p105 coactivator; SN4TDR; SND p102; SND1; snd1 protein; SND1-BRAF fusion; staphylococcal nuclease and tudor domain containing 1; staphylococcal nuclease domain containing 1; staphylococcal nuclease domain-containing protein 1; TDRD11; testis tissue sperm-binding protein Li 82P; tudor domain-containing protein 11; TudorSN; Tudor-SN; wu:fc27g10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27044 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
0.74 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q7KZF4 | |
Snd1 | |
Recombinant protein encompassing a sequence within the center region of human SND1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |