Läs mer
Ansell Edmont™ Snorkel™ 4-412 Series Green PVC Immersion Gloves
Handla allt Ansell Produkter
:
Size: 9
Beskrivning
- Chemical gloves
- Unique rough finished with wet-grip performance
- Jersey lining provide comfort and moderate insulation
- Good cut and abrasion performance
- For confident handling, especially in wet conditions
- Withstands abrasion three times longer than ordinary PVC-coated gloves
- Natural fit for improved dexterity and comfort
Applications:
- Filling, blending and charging of raw materials
- Loading and unloading process equipment
- Opening and draining pumps valves or lines
- Supervision of running operation
- Transferring liquids and solids
- Transferring liquids and solids between vessels and tanks and process equipment
Specifikationer
Specifikationer
| Material på baksidan | PVC |
| Certifikat och efterlevnad | Standards CATEGORY III, EN 374, EN 388 : 3121, ANSI |
| Manschettmaterial | PVC |
| Beläggningsmaterial | PVC |
| Användning av laboratorium/renrum | Yes |
| Produkttyp | Immersion Gloves |
| Stil | 4-412 |
| Handske manschettstil | Gauntlet |
| Foder | Cotton Jersey |
| Kvantitet | 72 Pairs |
| Visa mer |