Learn More
SNRNP40 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555589
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GDCDNYATLK GHSGAVMELH YNTDGSMLFS ASTDKTVAVW DSETGERVKR LKGHTSFVNS CYPARRGPQL VCTGSDDGTV KLWDIRKKAA IQ Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.
SNRNP40 Polyclonal antibody specifically detects SNRNP40 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
SNRNP40 | |
Polyclonal | |
Unconjugated | |
SNRNP40 | |
0610009C03Rik; 38 kDa-splicing factor; 40K; HPRP8BP; Prp8-binding protein; Prp8bp; PRPF8BP; RGD1309198; RP11-490K7.3; SFP38; small nuclear ribonucleoprotein 40 (U5); small nuclear ribonucleoprotein 40kDa (U5); small nuclear ribonucleoprotein U5 subunit 40; small nuclear ribonucleoprotein, U5 40kDa subunit; Snrnp40; SPF38; U5 small nuclear ribonucleoprotein 40 kDa protein; U5 snRNP 40 kDa protein; U5 snRNP-specific 40 kDa protein; U5 snRNP-specific 40 kDa protein (hPrp8-binding); U5 snRNP-specific protein (Prp8-binding); U5-40K; U5-40kD protein; WD repeat domain 57 (U5 snRNP specific); WD repeat-containing protein 57; Wdr57 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9410 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96DI7 | |
SNRNP40 | |
Recombinant protein corresponding to Human SNRNP40 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only