Learn More
SNRNP70 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541728
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PHNDPNAQGD AFKTLFVARV NYDTTESKLR REFEVYGPIK RIHMVYSKRS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
SNRNP70 Polyclonal antibody specifically detects SNRNP70 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
SNRNP70 | |
Polyclonal | |
Unconjugated | |
SNRNP70 | |
2700022N21Rik; 3200002N22Rik; AI325098; LOW QUALITY PROTEIN: U1 small nuclear ribonucleoprotein 70 kDa; R74807; RNPU1Z; Rnu1p70; Rnulp70; RPU1; small nuclear ribonucleoprotein 70 (U1); small nuclear ribonucleoprotein 70kDa; small nuclear ribonucleoprotein 70kDa (U1); small nuclear ribonucleoprotein 70kDa polypeptide (RNP antigen); small nuclear ribonucleoprotein U1 subunit 70; small nuclear ribonucleoprotein, U1 70kDa subunit; Snp1; Snrnp70; SNRP70; snrp70 protein; Srnp70; U1 small nuclear ribonucleoprotein 70 kDa; U1 small nuclear ribonucleoprotein polypeptide A; U1 small ribonucleoprotein 70kd; U1 snRNP 70 kDa; U1-70; U170K; U1-70K; U1AP; U1AP1; U1RNP; Unknown (protein for MGC:138058); wu:fc20b04; zgc:136450 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6625 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P08621 | |
SNRNP70 | |
Synthetic peptide directed towards the N-terminal of human SNRP70 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |