Learn More
SNRPA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561243
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: IIAKMKGTFV ERDRKREKRK PKSQETPATK KAVQGGGATP VVGAVQGPVP GMPPMTQAPR IMHHMPGQPP YMP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 95%.
The protein encoded by this gene associates with stem loop II of the U1 small nuclear ribonucleoprotein, which binds the 5' splice site of precursor mRNAs and is required for splicing. The encoded protein autoregulates itself by polyadenylation inhibition of its own pre-mRNA via dimerization and has been implicated in the coupling of splicing and polyadenylation.Specifications
SNRPA | |
Polyclonal | |
Unconjugated | |
SNRPA | |
C430021M15Rik; fc19d01; MGC137110 protein; Mud1; nRNP A; nuclear ribonucleoprotein A; protein A of the U1 small nuclear RNA; Rnu1a1; Rnu1a-1; small nuclear ribonucleoprotein polypeptide A; Snrpa; U1 small nuclear ribonucleoprotein A; U1 small nuclear RNP-specific A; U1 snRNP A; U1 snRNP protein A; U1 snRNP-specific protein A; U1A; U1-A; Unknown (protein for MGC:77810); wu:fc19d01; zgc:77810 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6626 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09012 | |
SNRPA | |
Recombinant protein corresponding to Human SNRPA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only