Learn More
SNRPG Recombinant Protein, Abnova
Brand:  Abnova H00006637-P01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16130892
  • 5030.41 SEK / 25µg
Estimated Shipment: 02-12-2021
Description
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
AAH00070
Solution
6637
SNRPG (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SNRPG
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
34.1
8
Glutathione Sepharose 4 Fast Flow
25 ug
MSKAHPPELKKFMDKKLSLKLNGGRHVQGILRGFDPFMNLVIDECVEMATSGQQNNIGMVVIRGNSIIMLEALERV
MGC117317/SMG
SNRPG