Learn More
SNX21 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561226
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RAQSLTCTGL YREALALWAN AWQLQAQLGT PSGPDRPLLT LAGLAVCHQE LEDPGEARAC CEKALQLLGD KSLHPLLAPF LEAHVRLSWR LGLDKRQSEA Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
SNX21 Polyclonal antibody specifically detects SNX21 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
SNX21 | |
Polyclonal | |
Unconjugated | |
SNX21 | |
C20orf161; dJ337O18.4; PP3993; SNX21; SNXL; SNX-L; sorting nexin 21; sorting nexin family member 21; sorting nexin L; Sorting nexin-21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90203 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969T3 | |
SNX21 | |
Recombinant protein corresponding to Human SNX21 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only