Learn More
Thermo Scientific Alfa Aesar 100ML SOB medium, contains 20g/L tryptone, 5g/L yeast extract, 2.4g/L MgSO4, 0.5g/L NaCl, and 0.186g
Brand:  Thermo Scientific Alfa Aesar J62210.AE
100ML SOB medium, contains 20g/L tryptone, 5g/L yeast extract, 2.4g/L MgSO4, 0.5g/L NaCl, and 0.186g
Additional Details : Weight : 0.00010kg
Product Code. 15484619