missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
SOD1/Cu-Zn SOD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20534
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
SOD1/Cu-Zn SOD Polyclonal antibody specifically detects SOD1/Cu-Zn SOD in Human, Mouse, Rat, Alligator, Avian samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifikationer
| SOD1/Cu-Zn SOD | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7), 20% Glycerol, 1% BSA with 0.025% Proclin 300 | |
| ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic | |
| Rabbit | |
| 16 kDa | |
| 0.1 mL | |
| Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors | |
| 6647 | |
| Human, Mouse, Rat, Alligator, Avian | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.29mg/mL | |
| Western Blot 1:500-1:10000, ELISA 1:1000-1:10000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen, Sandwich ELISA | |
| P00441 | |
| SOD1 | |
| Recombinant protein encompassing a sequence within the center region of human SOD1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat. Species reactivity based on sequence identity: Chimpanzee (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering