Learn More
SOD1 Mouse anti-Human, Unconjugated, Clone: 2F10G1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67480-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The SOD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SOD1. This antibody recognizes human, mouse, rat, pig antigen. The SOD1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
SOD1 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry, Immunofluorescence | |
2F10G1 | |
Unconjugated | |
20 μL | |
6647 | |
IgG2a |