missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten Graf™ Soda Lime Glass Class A Pipets
Brand: Poulten Graf™ R371/WAC/E/5
Additional Details : Weight : 0.29520kg
Specifications
Yes | |
A | |
2 Pack | |
5 mL |
To Deliver | |
Soda Lime Glass | |
0.015 mL | |
Bulb Pipet |