Läs mer
Sodium 1,6,6-Trimethyl-10,11-dioxo-6,7,8,9,10,11-hexahydrophenanthro[1,2-b]furan-2-sulfonate, TRC
Produktkod. 30424478
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30424478 25 mg
1 options
Produktkod. 30424478 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS67378525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sodium 1,6,6-Trimethyl-10,11-dioxo-6,7,8,9,10,11-hexahydrophenanthro[1,2-b]furan-2-sulfonate
CAS 69659-80-9
Molekylformel C19H17NaO6S
Kvantitet 25 mg
Synonym 6,7,8,9,10,11-Hexahydro-1,6,6-trimethyl-10,11-dioxo-phenanthro[1,2-b]furan-2-sulfonic Acid Sodium Salt (1:1), DS 201, Sodium Tanshinone IIA Sulfonate, Sulfotanshinone Sodium II-A, Tanshinone IIA Sodium Sulfonate
InChI formel InChI=1S/C19H18O6S.Na/c1-9-13-15(20)16(21)14-10-5-4-8-19(2,3)12(10)7-6-11(14)17(13)25-18(9)26(22,23)24;/h6-7H,4-5,8H2,1-3H3,(H,22,23,24);/q;+1/p-1
LEDER [Na+].Cc1c2C(=O)C(=O)c3c4CCCC(C)(C)c4ccc3c2oc1S(=O)(=O)[O-]
IUPAC-namn sodium;1,6,6-trimethyl-10,11-dioxo-8,9-dihydro-7H-naphtho[1,2-g][1]benzofuran-2-sulfonate
Molekylvikt (g/mol) 396.39
Formel vikt 396.0644