Läs mer
Sodium 2-[2-[(2-Bromo-6-chlorophenyl)amino]phenyl]acetate, Mikromol
Produktkod. 30461372
Change view
Kvantitet:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461372 4 x 25 mg
30050746 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30461372 Leverantör Mikromol Leverantörsnummer MM0006.0500254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Sodium 2-[2-[(2-Bromo-6-chlorophenyl)amino]phenyl]acetate, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Sodium 2-[2-[(2-Bromo-6-chlorophenyl)amino]phenyl]acetate
CAS 127792-45-4
Analyt- eller komponentnamn Sodium 2-[2-[(2-Bromo-6-chlorophenyl)amino]phenyl]acetate
Frakt skick Room Temperature
Molekylformel C14 H10 Br Cl N O2 . Na
Certifikat och efterlevnad ISO 9001
Synonym Diclofenac Sodium Imp. D (EP), Diclofenac Imp. D (EP), Sodium 2-[2-[(2-Bromo-6-chlorophenyl)amino]phenyl]acetate, Diclofenac Sodium Imp. D (EP) as Sodium Salt, Diclofenac Sodium Impurity D as Sodium Salt, Diclofenac Impurity D as Sodium Salt
InChI formel InChI=1S/C14H11BrClNO2.Na/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;/h1-7,17H,8H2,(H,18,19);/q;+1/p-1
Lösningstyp Neat
LEDER [Na+].[O-]C(=O)Cc1ccccc1Nc2c(Cl)cccc2Br
IUPAC-namn sodium;2-[2-(2-bromo-6-chloroanilino)phenyl]acetate
Molekylvikt (g/mol) 362.58
Kvantitet 4 x 25 mg
Formel vikt 360.9481 g/mol