Läs mer
Sodium 2-Ethylhexyl Sulfate (40%-60% in Water), TRC
Produktkod. 30009497
Change view
Kvantitet:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009497 5 g
30011027 50 g
30006462 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009497 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6994055G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 126-92-1
Molekylformel C8H17NaO4S
Molekylvikt (g/mol) 232.27
Synonym 2-Ethylhexyl Sulfate Sodium Salt, Avirol SA 4106, Carsonol SHS, Disponil EHS 47, Emcol D 5-10, Ethasulfate Sodium, Kraftex OA, Lugalvan TC-EHS, Lutensit TC-EHS, NAS 08, NSC 4744, Newcol 1000SN, Niaproof 08, Nissan Sintrex EHR, Pentrone ON, Pionin A 20, Rewopol NEHS 40, Rhodapon BOS, Rhodapon OLS, Sandet OHE, Sinolin SO 35, Sintrex EHR, Sodium 2-ethylhexyl Sulfate, Sodium Etasulfate, Sodium Ethasulfate, Sodium Octyl Sulfate, Sodium Octyl Sulphate, Sole Tege TS 25, Stepanol EHS, Steponol EHS, Sulfirol 8, Sulfopon O, Sulfotex CA, Sulfotex OA, Supralate SP, TC-EHS, Tergemist, Tergimist, Tergitol 08, Tergitol Anionic 08, Texapon 890, Texapon EHS, Witcolate D 5-10
IUPAC-namn sodium;2-ethylhexyl sulfate
LEDER [Na+].CCCCC(CC)COS(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material Sodium 2-Ethylhexyl Sulfate
Kvantitet 5 g
InChI formel InChI=1S/C8H18O4S.Na/c1-3-5-6-8(4-2)7-12-13(9,10)11;/h8H,3-7H2,1-2H3,(H,9,10,11);/q;+1/p-1
Formel vikt 232.0745
Koncentration eller sammansättning (efter analyt eller komponenter) Sodium 2-Ethylhexyl Sulfate 40%-60%